1,2-Dimethylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dimethyl-1H-benzo[d]imidazole

A435300-100MG A435300-250MG A435300-5G
77116-110EA 27.59 USD
77116-110 77116-112 77116-114 77116-116
1,2-Dimethylbenzimidazole 98%
1,2-Dimethylbenzimidazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD00022840
CAS Number: 2876-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR