3,5-Difluoro-2-methoxybenzyl bromide 95%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-3,5-difluoro-2-methoxybenzene

A435276-10G A435276-1G A435276-250MG A435276-5G
77116-102EA 178.24 USD
77116-102 77116-104 77116-106 77116-108
3,5-Difluoro-2-methoxybenzyl bromide 95%
3,5-Difluoro-2-methoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
MDL Number: MFCD06660240
CAS Number: 886500-63-6
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR