4-Chloro-3-(chlorosulfonyl)benzoic acid 95%
Supplier: Ambeed
|
Danger
|
A434231-1G
A434231-5G
77116-096EA
9.27
USD
77116-096
77116-098
4-Chloro-3-(chlorosulfonyl)benzoic acid 95%
4-Chloro-3-(chlorosulfonyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄Cl₂O₄S |
MDL Number:
MFCD00625723 CAS Number: 2494-79-3 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...