3-Amino-3-(3-methylphenyl)propionic acid 98%

Supplier: Ambeed
Warning

A434105-1G A434105-250MG A434105-25G A434105-5G
77116-092EA 10.31 USD
77116-092 77116-094 77686-181 77686-182
3-Amino-3-(3-methylphenyl)propionic acid 98%
3-Amino-3-(3-methylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD01871312
CAS Number: 68208-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR