1-Boc-piperazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

A433625-10G A433625-1G A433625-250MG A433625-25G
77116-074EA 200.52 USD
77116-074 77116-076 77116-078 77116-080
1-Boc-piperazine-2-carboxylic acid 97%
1-Boc-piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.2600 g/mol
MDL Number: MFCD02682403
CAS Number: 1214196-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR