Neotame 99% (contains < 5% H₂O)

Supplier: Ambeed
Warning

Synonyms: (S)-3-((3,3-Dimethylbutyl)amino)-4-(((S)-1-methoxy-1-oxo-3-phenylpropan-2-yl)amino)-4-oxobutanoic acid, N-[N-(3,3-Dimethylbutyl)-L-alpha-aspartyl]-L-phenylalanine 1-Methyl ester, N-(N-(3,3-Dimethylbutyl)-L-alpha-aspartyl)-L-phenylalanine 1-methyl ester

A431326-1000G A431326-100G A431326-10G A431326-25G A431326-500G A431326-5G
77116-052EA 258.86 USD
77116-052 77116-054 77116-056 77116-058 77116-060 77116-062
Neotame 99% (contains < 5% H₂O)
Neotame

Precaución: for Research Use Only

Formula: C₂₀H₃₀N₂O₅
MW: 378.47 g/mol
MDL Number: MFCD09039056
CAS Number: 165450-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR