Methyl-6-amino-5-bromonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-amino-5-bromonicotinate

A431293-1G A431293-250MG A431293-5G A431293-25G
77116-046EA 19.1 USD
77116-046 77116-048 77116-050 77686-175
Methyl-6-amino-5-bromonicotinate 97%
Methyl-6-amino-5-bromonicotinate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR