1-Methoxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-naphthyl ether

A431272-1000G A431272-100G A431272-10G A431272-25G A431272-500G
77116-036EA 149.59 USD
77116-036 77116-038 77116-040 77116-042 77116-044
1-Methoxynaphthalene 98%
1-Methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.2 g/mol
MDL Number: MFCD00003924
CAS Number: 2216-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR