4-(Hydroxymethyl)tetrahydro-2H-pyran-4-carbonitrile 98%

Supplier: Ambeed
Danger

A430107-100MG A430107-250MG
77116-032EA 54.11 USD
77116-032 77116-034
4-(Hydroxymethyl)tetrahydro-2H-pyran-4-carbonitrile 98%
4-(Hydroxymethyl)tetrahydro-2H-pyran-4-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂
MW: 141.16 g/mol
MDL Number: MFCD19103514
CAS Number: 1010836-56-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR