5-(Methylthio)thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylsulphanylthiophene-2-carboxylic acid

A429410-1G A429410-250MG
77116-026EA 124.13 USD
77116-026 77116-028
5-(Methylthio)thiophene-2-carboxylic acid 98%
5-(Methylthio)thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆O₂S₂
MW: 174.232 g/mol
MDL Number: MFCD00173742
CAS Number: 20873-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR