2,4-Dichloro-5-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A428734-100G A428734-1G A428734-25G A428734-5G
77116-012EA 399.96 USD
77116-012 77116-014 77116-016 77116-018
2,4-Dichloro-5-fluorobenzoic acid 98%
2,4-Dichloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
MW: 209 g/mol
MDL Number: MFCD00075281
CAS Number: 86522-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR