3-Chloro-2,6-difluorobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A426742-10G
A426742-1G
A426742-250MG
A426742-25G
A426742-5G
77115-984EA
70.02
USD
77115-984
77115-986
77115-988
77115-990
77115-992
3-Chloro-2,6-difluorobenzoic acid 97%
3-Chloro-2,6-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃ClF₂O₂ MW: 192.55 g/mol |
MDL Number:
MFCD01631322 CAS Number: 225104-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...