4-Chloro-2-fluoro-3-methylbenzonitrile 95%

Supplier: Ambeed
Danger

A425197-100MG A425197-1G A425197-250MG
77115-972EA 16.47 USD
77115-972 77115-974 77115-976
4-Chloro-2-fluoro-3-methylbenzonitrile 95%
4-Chloro-2-fluoro-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MDL Number: MFCD28751902
CAS Number: 1207875-88-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR