2-Fluorophenol 98%

Supplier: Ambeed
Danger

A423211-10G A423211-25G A423211-100G
77115-968EA 16.46 USD
77115-968 77115-970 77686-163
2-Fluorophenol 98%
2-Fluorophenol

Precaución: for Research Use Only

Formula: C₆H₅FO
MDL Number: MFCD00002155
CAS Number: 367-12-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR