O-Methylhydroxylamine hydrochloride 98%
Supplier: Ambeed
|
Danger
|
A422985-25G
A422985-100G
77115-966EA
5.15
USD
77115-966
77572-314
O-Methylhydroxylamine hydrochloride 98%
O-Methylhydroxylamine hydrochloride
Precaución: for Research Use Only
Formula:
CH₆ClNO |
MDL Number:
MFCD00012951 CAS Number: 593-56-6 UN: 1759 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...