2-Bromo-4-chloro-6-nitroaniline 95%
Supplier: Ambeed
|
Warning
|
A422633-100G
A422633-10G
A422633-1G
A422633-25G
A422633-500G
A422633-5G
77115-954EA
64.88
USD
77115-954
77115-956
77115-958
77115-960
77115-962
77115-964
2-Bromo-4-chloro-6-nitroaniline 95%
2-Bromo-4-chloro-6-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrClN₂O₂ |
MDL Number:
MFCD08275709 CAS Number: 827-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...