(-)-Arctigenin 98%

Supplier: Ambeed
Warning

A420992-100MG A420992-10MG A420992-1G A420992-1MG A420992-250MG A420992-50MG A420992-5G
77115-924EA 32.96 USD
77115-924 77115-926 77115-928 77115-930 77115-932 77115-934 77613-296
(-)-Arctigenin 98%
(-)-Arctigenin

Precaución: for Research Use Only

Formula: C₂₁H₂₄O₆
MDL Number: MFCD00870597
CAS Number: 7770-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR