2-(4-Amino-2-nitrophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A419121-1G A419121-250MG A419121-5G
77115-918EA 313.12 USD
77115-918 77115-920 77115-922
2-(4-Amino-2-nitrophenyl)acetic acid 95%
2-(4-Amino-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MDL Number: MFCD01110332
CAS Number: 116435-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR