2,4-Difluoropyridine 98%

Supplier: Ambeed
Danger

A416126-10G A416126-1G A416126-25G A416126-5G
77115-890EA 23.69 USD
77115-890 77115-892 77115-894 77115-896
2,4-Difluoropyridine 98%
2,4-Difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃F₂N
MDL Number: MFCD04114162
CAS Number: 34941-90-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR