4,5-Diamino-6-hydroxypyrimidine 95%

Supplier: Ambeed
Warning

A416122-10G A416122-250MG A416122-25G A416122-5G A416122-1G
77115-882EA 92.71 USD
77115-882 77115-884 77115-886 77115-888 77686-147
4,5-Diamino-6-hydroxypyrimidine 95%
4,5-Diamino-6-hydroxypyrimidine

Precaución: for Research Use Only

Formula: C₄H₆N₄O
MDL Number: MFCD00006114
CAS Number: 1672-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR