3-Bromo-6-nitro-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-nitroindazole

A411878-250MG A411878-5G
77115-842EA 9.26 USD
77115-842 77115-844
3-Bromo-6-nitro-1H-indazole 98%
3-Bromo-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.0300 g/mol
MDL Number: MFCD06410881
CAS Number: 70315-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR