4-Phenylpiperidine hydrochloride 97%

Supplier: Ambeed
Warning

A411175-10G A411175-1G
77115-828EA 84.88 USD
77115-828 77115-830
4-Phenylpiperidine hydrochloride 97%
4-Phenylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClN
MW: 197.7044 g/mol
MDL Number: MFCD01632527
CAS Number: 10272-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR