Methyl-3-(chlorosulfonyl)benzoate 96%

Supplier: Ambeed
Danger

Synonyms: Methyl 3-(chlorosulphonyl)benzoate

A409263-10G A409263-1G A409263-25G A409263-5G
77115-812EA 46.35 USD
77115-812 77115-814 77115-816 77115-818
Methyl-3-(chlorosulfonyl)benzoate 96%
Methyl 3-(chlorosulphonyl)benzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₄S
MW: 234.6600 g/mol
MDL Number: MFCD06408800
CAS Number: 63555-50-0
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR