Fmoc-4-Abz-OH 97%

Supplier: Ambeed
Warning

A408990-100G A408990-25G A408990-5G
77115-798EA 298.12 USD
77115-798 77115-800 77115-802
Fmoc-4-Abz-OH 97%
Fmoc-4-Abz-OH

Precaución: for Research Use Only

Formula: C₂₂H₁₇NO₄
MW: 359.381 g/mol
MDL Number: MFCD00144888
CAS Number: 185116-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR