(R)-Methyl-2-amino-2-(4-hydroxyphenyl)acetate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride, (R)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride

A406872-100G A406872-10G A406872-25G A406872-5G
77115-768EA 42.22 USD
77115-768 77115-770 77115-772 77115-774
(R)-Methyl-2-amino-2-(4-hydroxyphenyl)acetate hydrochloride 95%
(R)-Methyl-2-amino-2-(4-hydroxyphenyl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₃
MDL Number: MFCD01862152
CAS Number: 57591-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR