2-Chloro-1,3-dimethylimidazolium tetrafluoroborate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-1,3-dimethylimidazolinium tetrafluoroborate, 2-Chloro-1,3-dimethyl-4,5-dihydroimidazolium tetrafluoroborate, CIB, 2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate
A405797-100G
A405797-10G
A405797-1G
A405797-25G
A405797-5G
77115-754EA
302.36
USD
77115-754
77115-756
77115-758
77115-760
77115-762
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate 98%
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₅H₁₀ClN₂·BF₄ MW: 220.4 g/mol |
MDL Number:
MFCD09039291 CAS Number: 153433-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...