(±)-2-Amino-2-(4-chlorophenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
DL-2-(4-Chlorophenyl)glycine, H-DL-Phg(4-Cl)-OH, DL-alpha-Amino-4-chlorophenylacetic acid, 4-Chloro-DL-2-phenylglycine, 4-Chloro-DL-phenylglycine, 2-Amino-2-(4-chlorophenyl)acetic acid
A400902-1000G
A400902-100G
A400902-25G
A400902-500G
A400902-5G
77115-726EA
244.12
USD
77115-726
77115-728
77115-730
77115-732
77115-734
77115-736
(±)-2-Amino-2-(4-chlorophenyl)acetic acid 97%
(±)-2-Amino-2-(4-chlorophenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₂ MW: 185.61 g/mol |
MDL Number:
MFCD00049328 CAS Number: 6212-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...