2,4-Dimethylbenzylamine 98%

Supplier: Ambeed
Danger

A400018-1G A400018-250MG A400018-25G A400018-5G
77115-708EA 60.48 USD
77115-708 77115-710 77115-712 77115-714
2,4-Dimethylbenzylamine 98%
2,4-Dimethylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00025575
CAS Number: 94-98-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR