4-Bromo-3-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A395646-100G A395646-10G A395646-1G A395646-25G
77115-684EA 405.27 USD
77115-684 77115-686 77115-688 77115-690
4-Bromo-3-nitrobenzaldehyde 97%
4-Bromo-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃
MW: 230.02 g/mol
MDL Number: MFCD00204031
CAS Number: 163596-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR