α-Bromo-2-chloro-6-fluorotoluene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Chloro-6-fluorobenzyl bromide, 2-(Bromomethyl)-1-chloro-3-fluorobenzene
A392833-10G
A392833-1G
A392833-25G
A392833-5G
77115-662EA
21.63
USD
77115-662
77115-664
77115-666
77115-668
α-Bromo-2-chloro-6-fluorotoluene 98%
α-Bromo-2-chloro-6-fluorotoluene
Precaución: for Research Use Only
Formula:
C₇H₅BrClF MW: 223.47 g/mol Boiling Pt: 223 Melting Pt: 37 |
MDL Number:
MFCD00040126 CAS Number: 68220-26-8 UN: 3265 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...