(3S,4S)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Danger

A389866-1G A389866-250MG
77115-626EA 466.8 USD
77115-626 77115-628
(3S,4S)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate 95%
(3S,4S)-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₇FN₂O₂
MW: 204.24 g/mol
MDL Number: MFCD11111576
CAS Number: 1009075-43-7
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR