2,4,6-Tribromo-3-hydroxybenzaldehyde 98+%

Supplier: Ambeed
Warning

A389318-100G A389318-1G A389318-25G A389318-5G
77115-612EA 560.16 USD
77115-612 77115-614 77115-616 77115-618
2,4,6-Tribromo-3-hydroxybenzaldehyde 98+%
2,4,6-Tribromo-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Br₃O₂
MW: 358.81 g/mol
MDL Number: MFCD00017320
CAS Number: 2737-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR