(S)-Pyrrolidine-2-carbonitrile hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Pyrrolidinecarbonitrile hydrochloride

A388821-100MG A388821-1G A388821-250MG A388821-5G
77115-606EA 42.44 USD
77115-606 77115-608 77115-610 77594-182
(S)-Pyrrolidine-2-carbonitrile hydrochloride 98%
(S)-Pyrrolidine-2-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂
MW: 132.5914 g/mol
MDL Number: MFCD02683060
CAS Number: 65732-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR