4-Chloro-2-methylbenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A387592-100G
A387592-10G
A387592-1G
A387592-25G
A387592-5G
77115-578EA
159.14
USD
77115-578
77115-580
77115-582
77115-584
77115-586
4-Chloro-2-methylbenzonitrile 98%
4-Chloro-2-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆ClN MW: 151.60 g/mol |
MDL Number:
MFCD00045603 CAS Number: 50712-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...