α-Bromo-2,6-difluorotoluene 97%

Supplier: Ambeed
Danger

A386878-10G A386878-1G A386878-25G A386878-5G
77115-570EA 17.51 USD
77115-570 77115-572 77115-574 77115-576
α-Bromo-2,6-difluorotoluene 97%
α-Bromo-2,6-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MDL Number: MFCD00000329
CAS Number: 85118-00-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR