3,4,5-Trifluoro-DL-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(3,4,5-trifluorophenyl)propanoic acid, H-DL-Phe(3,4,5-TriF)-OH

A386808-100MG A386808-1G A386808-250MG
77115-564EA 25.47 USD
77115-564 77115-566 77115-568
3,4,5-Trifluoro-DL-phenylalanine 95%
3,4,5-Trifluoro-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD01631414
CAS Number: 261952-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR