2-Propanesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: Isopropylsulphonyl chloride

A385458-10G A385458-1G A385458-5G A385458-25G
77115-554EA 37.08 USD
77115-554 77115-556 77115-558 77551-134
2-Propanesulphonyl chloride 98%
2-Propanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₃H₇ClO₂S
MW: 142.61 g/mol
Boiling Pt: 181
Melting Pt: -47
Flash Pt: 86
MDL Number: MFCD00007453
CAS Number: 10147-37-2
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR