(S)-1-N-Boc-4-N-Fmoc-piperazine2-carboxylic acid 97%

Supplier: Ambeed
Warning

A384932-100MG A384932-1G A384932-250MG A384932-5G
77115-546EA 16.98 USD
77115-546 77115-548 77115-550 77115-552
(S)-1-N-Boc-4-N-Fmoc-piperazine2-carboxylic acid 97%
(S)-1-N-Boc-4-N-Fmoc-piperazine2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₂O₆
MW: 452.50 g/mol
MDL Number: MFCD02179101
CAS Number: 1034574-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR