3-Amino-6-chloropyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

A383744-1G A383744-250MG
77115-534EA 103.97 USD
77115-534 77115-536
3-Amino-6-chloropyridine-2-carbonitrile 98%
3-Amino-6-chloropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MDL Number: MFCD07780767
CAS Number: 95095-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR