3-Hydroxy-2-nitro-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-2-nitropyridin-3-ol, 6-Methyl-2-nitro-3-pyridone, 3-Hydroxy-6-methyl-2-nitropyridine, 6-Metil-2-nitropyridine-3-ol

A383338-10G A383338-1G A383338-25G A383338-5G
77115-520EA 57.29 USD
77115-520 77115-522 77115-524 77115-526
3-Hydroxy-2-nitro-6-picoline 98%
3-Hydroxy-2-nitro-6-picoline

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00006259
CAS Number: 15128-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR