2-Amino-3-bromo-5,6-dimethylpyridine 98%

Supplier: Ambeed
Warning

A383212-1G A383212-250MG A383212-5G
77115-516EA 48.41 USD
77115-516 77115-518 77551-114
2-Amino-3-bromo-5,6-dimethylpyridine 98%
2-Amino-3-bromo-5,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD04039386
CAS Number: 161091-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR