5-Amino-2,3-dichloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichloropyridin-5-amine, 2,3-Dichloro-5-pyridylamine, 3-Amino-5,6-dichloropyridine

A380938-100G A380938-250MG A380938-25G A380938-5G
77115-502EA 543.19 USD
77115-502 77115-504 77115-506 77115-508
5-Amino-2,3-dichloropyridine 98%
5-Amino-2,3-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
MDL Number: MFCD03840434
CAS Number: 98121-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR