N-Biphenyl-4-yl-N',N'-diphenylbenzene-1,4-diamine 97%

Supplier: Ambeed
Warning

A379405-100MG A379405-1G A379405-250MG
77115-484EA 20.16 USD
77115-484 77115-486 77115-488
N-Biphenyl-4-yl-N',N'-diphenylbenzene-1,4-diamine 97%
N-Biphenyl-4-yl-N',N'-diphenylbenzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₃₀H₂₄N₂
MW: 412.52 g/mol
MDL Number: MFCD09878766
CAS Number: 880800-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR