3,4-Difluorobenzoyl chloride 99%

Supplier: Ambeed
Danger

Synonyms: 3,4-difluorobenzoylchloride

A378696-25G A378696-5G
77115-480EA 64.88 USD
77115-480 77115-482
3,4-Difluorobenzoyl chloride 99%
3,4-Difluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
MDL Number: MFCD00009930
CAS Number: 76903-88-3
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR