3-Amino-2-chloro-4-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine
A376397-1000G
A376397-100G
A376397-10G
A376397-25G
A376397-500G
A376397-5G
77115-450EA
305.91
USD
77115-450
77115-452
77115-454
77115-456
77115-458
77686-054
3-Amino-2-chloro-4-methylpyridine 98%
3-Amino-2-chloro-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂ |
MDL Number:
MFCD00673152 CAS Number: 133627-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...