2,6-Dichloro-4-methylbenzaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichloro-p-tolualdehyde, 3,5-Dichloro-4-formyltoluene
A374341-250MG
A374341-5G
A374341-1G
77115-420EA
57.29
USD
77115-420
77115-422
77594-122
2,6-Dichloro-4-methylbenzaldehyde 95%
2,6-Dichloro-4-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆Cl₂O MW: 189.039 g/mol |
MDL Number:
MFCD09753749 CAS Number: 116070-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...