2,3-Dichloro-5-pyridineboronic acid 97%

Supplier: Ambeed
Warning

A372845-250MG A372845-5G
77115-414EA 31.93 USD
77115-414 77115-416
2,3-Dichloro-5-pyridineboronic acid 97%
2,3-Dichloro-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
MDL Number: MFCD08277308
CAS Number: 1072944-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR