SNX-2112 ≥98%

Supplier: Ambeed
Warning

Synonyms: PF-04928473, 2-[(trans-4-Hydroxycyclohexyl)amino]-4-[4,5,6,7-tetrahydro-6,6-dimethyl-4-oxo-3-(trifluoromethyl)-1H-indazol-1-yl]benzamide, 4-(6,6-Dimethyl-4-oxo-3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl)-2-((trans-4-hydroxycyclohexyl)amino)benzamide

A371641-10MG A371641-5MG
77115-410EA 70.02 USD
77115-410 77115-412
SNX-2112 ≥98%
SNX-2112

Precaución: for Research Use Only

Formula: C₂₃H₂₇F₃N₄O₃
MW: 464.48 g/mol
MDL Number: MFCD16038907
CAS Number: 908112-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR