Ethyl (4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

A370976-100MG A370976-1G A370976-250MG
77115-404EA 63.66 USD
77115-404 77115-406 77115-408
Ethyl (4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate 95%
Ethyl (4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₁₀BrN₃O₄
MW: 292.1 g/mol
MDL Number: MFCD00682561
CAS Number: 351992-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR