Z-D-3-Abu-OH 97%

Supplier: Ambeed
Warning

Synonyms: Z-β-D-HomoAla-OH

A370128-100G A370128-1G A370128-250MG A370128-25G A370128-5G
77115-394EA 962.24 USD
77115-394 77115-396 77115-398 77115-400 77115-402
Z-D-3-Abu-OH 97%
Z-D-3-Abu-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
MDL Number: MFCD06223032
CAS Number: 67843-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR